Assay Details
| Gene Symbol: LOC108638200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063972.1 | XP_017919461.1 | 1 - 2 | 191 | 75 | |
| Gene Symbol: PRKCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063971.1 | XP_017919460.1 | 1717 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018063972.1 transcripts.
Set Membership: |
|

LOC108638200
Genomic Map