Assay Details
| Gene Symbol: LOC108638200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063972.1 | XP_017919461.1 | 2 - 3 | 297 | 96 | |
| Gene Symbol: PRKCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063971.1 | XP_017919460.1 | 1823 | 96 | ||
More Information
Set Membership: |
|

LOC108638200
Genomic Map