Assay Details
| Gene Symbol: MAPK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038788.1 | XP_017894277.1 | 9 - 10 | 846 | 60 | |
| GenBank mRNA | JI861647.1 | - | 9 - 10 | 757 | 60 | |
| JP351361.2 | - | 9 - 10 | 873 | 60 | ||
| KA324986.1 | - | 3 - 4 | 698 | 60 | ||
More Information
Set Membership: |
|

MAPK13
Genomic Map