Assay Details
| Gene Symbol: SLC9A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043843.1 | XP_017899332.1 | 8 - 9 | 1282 | 59 | |
| XM_018043844.1 | XP_017899333.1 | 8 - 9 | 1282 | 59 | ||
| GenBank mRNA | JO604031.2 | - | 8 - 9 | 1214 | 59 | |
More Information
Set Membership: |
|

SLC9A7
Genomic Map