Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674841.3 | XP_005674898.3 | 9 - 10 | 1968 | 67 | |
| GenBank mRNA | JO335317.2 | - | 3 - 4 | 620 | 67 | |
| JO335318.2 | - | 3 - 4 | 620 | 67 | ||
| JO366392.2 | - | 3 - 4 | 740 | 67 | ||
| JO366393.2 | - | 3 - 4 | 740 | 67 | ||
| JO377585.2 | - | 3 - 4 | 795 | 67 | ||
| JO428097.1 | - | 3 - 4 | 899 | 67 | ||
| JO439836.2 | - | 3 - 4 | 839 | 67 | ||
| JO498420.2 | - | 3 - 4 | 704 | 67 | ||
| JO506906.2 | - | 3 - 4 | 704 | 67 | ||
| JO542650.2 | - | 3 - 4 | 312 | 67 | ||
| JO542651.2 | - | 3 - 4 | 312 | 67 | ||
| JO542652.2 | - | 3 - 4 | 312 | 67 | ||
| JO559669.2 | - | 9 - 10 | 1316 | 67 | ||
| JO560830.2 | - | 9 - 10 | 1316 | 67 | ||
| JO576572.1 | - | 3 - 4 | 713 | 67 | ||
| JP351338.2 | - | 9 - 10 | 1313 | 67 | ||
| JT658318.1 | - | 6 - 7 | 967 | 67 | ||
| JT763182.1 | - | 9 - 10 | 1180 | 67 | ||
| KA342503.1 | - | 9 - 10 | 1679 | 67 | ||
More Information
Set Membership: |
|

TOMM70
Genomic Map