Assay Details
| Gene Symbol: C3H1orf226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046419.1 | XP_017901908.1 | 10 - 11 | 1503 | 110 | |
| XM_018046420.1 | XP_017901909.1 | 10 - 11 | 1488 | 110 | ||
More Information
Set Membership: |
|

C3H1orf226
Genomic Map