Assay Details
| Gene Symbol: RHNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048634.1 | XP_017904123.1 | 2 - 3 | 292 | 103 | |
| XM_018048635.1 | XP_017904124.1 | 2 - 3 | 337 | 103 | ||
More Information
Set Membership: |
|

RHNO1
Genomic Map