Assay Details
| Gene Symbol: LECT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682961.3 | XP_005683018.1 | 1 - 2 | 279 | 72 | |
| GenBank mRNA | JO421480.2 | - | 3 - 4 | 685 | 72 | |
More Information
Set Membership: |
|

LECT2
Genomic Map