Assay Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684173.2 | XP_005684230.2 | 2 - 3 | 563 | 65 | |
| XM_018052499.1 | XP_017907988.1 | 2 - 3 | 572 | 65 | ||
| GenBank mRNA | KA345176.1 | - | 2 - 3 | 344 | 65 | |
More Information
Set Membership: |
|

CARD19
Genomic Map