Assay Details
| Gene Symbol: TSPYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684541.3 | XP_005684598.2 | 1 | 876 | 107 | |
| GenBank mRNA | JO403800.2 | - | 1 | 1301 | 107 | |
| JO425721.1 | - | 2 | 2814 | 107 | ||
More Information
Set Membership: |
|

TSPYL1
Genomic Map