Assay Details
| Gene Symbol: LOC102175216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054405.1 | XP_017909894.1 | 7 - 8 | 813 | 71 | |
| XM_018054406.1 | XP_017909895.1 | 7 - 8 | 807 | 71 | ||
| XM_018054407.1 | XP_017909896.1 | 6 - 7 | 1294 | 71 | ||
| XM_018054408.1 | XP_017909897.1 | 8 - 9 | 699 | 71 | ||
| XM_018054409.1 | XP_017909898.1 | 7 - 8 | 1180 | 71 | ||
| XM_018054411.1 | XP_017909900.1 | 12 - 13 | 1220 | 71 | ||
More Information
Set Membership: |
|

LOC102175216
Genomic Map