Assay Details
| Gene Symbol: LOC102175216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054405.1 | XP_017909894.1 | 9 - 10 | 1025 | 92 | |
| XM_018054406.1 | XP_017909895.1 | 9 - 10 | 1019 | 92 | ||
| XM_018054407.1 | XP_017909896.1 | 8 - 9 | 1506 | 92 | ||
| XM_018054408.1 | XP_017909897.1 | 10 - 11 | 911 | 92 | ||
| XM_018054409.1 | XP_017909898.1 | 9 - 10 | 1392 | 92 | ||
| XM_018054411.1 | XP_017909900.1 | 14 - 15 | 1432 | 92 | ||
More Information
Set Membership: |
|

LOC102175216
Genomic Map