Assay Details
| Gene Symbol: LOC102175216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054405.1 | XP_017909894.1 | 15 - 16 | 1822 | 66 | |
| XM_018054406.1 | XP_017909895.1 | 15 - 16 | 1816 | 66 | ||
| XM_018054407.1 | XP_017909896.1 | 14 - 15 | 2303 | 66 | ||
| XM_018054408.1 | XP_017909897.1 | 16 - 17 | 1708 | 66 | ||
| XM_018054409.1 | XP_017909898.1 | 15 - 16 | 2189 | 66 | ||
| XM_018054411.1 | XP_017909900.1 | 20 - 21 | 2229 | 66 | ||
| GenBank mRNA | JR119638.1 | - | 395 | 66 | ||
More Information
Set Membership: |
|

LOC102175216
Genomic Map