Assay Details
| Gene Symbol: MYZAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685718.3 | XP_005685775.2 | 9 - 10 | 1065 | 78 | |
| XM_013967112.2 | XP_013822566.2 | 9 - 10 | 1077 | 78 | ||
More Information
Set Membership: |
|

MYZAP
Genomic Map