Assay Details
| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685795.3 | XP_005685852.2 | 5 - 6 | 756 | 60 | |
| XM_005685796.3 | XP_005685853.2 | 5 - 6 | 714 | 60 | ||
| XM_013967150.2 | XP_013822604.2 | 5 - 6 | 547 | 60 | ||
| XM_018054176.1 | XP_017909665.1 | 4 - 5 | 565 | 60 | ||
| GenBank mRNA | JO598540.2 | - | 5 - 6 | 626 | 60 | |
More Information
Set Membership: |
|

GALK2
Genomic Map