Assay Details
| Gene Symbol: ITGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056382.1 | XP_017911871.1 | 8 - 9 | 1381 | 63 | |
| GenBank mRNA | JO556874.2 | - | 8 - 9 | 1392 | 63 | |
| KA334680.1 | - | 3 - 4 | 1124 | 63 | ||
More Information
Set Membership: |
|

ITGBL1
Genomic Map