Assay Details
| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688650.3 | XP_005688707.1 | 3 - 4 | 1928 | 67 | |
| XM_018058030.1 | XP_017913519.1 | 4 - 5 | 553 | 67 | ||
| XM_018058031.1 | XP_017913520.1 | 4 - 5 | 1308 | 67 | ||
| XM_018058032.1 | XP_017913521.1 | 4 - 5 | 2008 | 67 | ||
| XM_018058033.1 | XP_017913522.1 | 2 - 3 | 221 | 67 | ||
| XM_018058034.1 | XP_017913523.1 | 3 - 4 | 373 | 67 | ||
| XM_018058035.1 | XP_017913524.1 | 3 - 4 | 1230 | 67 | ||
| XM_018058036.1 | XP_017913525.1 | 3 - 4 | 452 | 67 | ||
More Information
Set Membership: |
|

ADNP
Genomic Map