Assay Details
| Gene Symbol: RGS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690948.3 | XP_005691005.3 | 2 - 3 | 430 | 77 | |
| GenBank mRNA | JO635359.2 | - | 2 - 3 | 273 | 77 | |
More Information
Set Membership: |
|

RGS16
Genomic Map