Assay Details
| Gene Symbol: PSMD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691403.3 | XP_005691460.1 | 2 - 3 | 353 | 61 | |
| XM_005691404.3 | XP_005691461.1 | 2 - 3 | 355 | 61 | ||
| GenBank mRNA | JO612585.2 | - | 2 - 3 | 280 | 61 | |
| JR138825.1 | - | 1280 | 61 | |||
More Information
Set Membership: |
|

PSMD9
Genomic Map