Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695598.3 | XP_005695655.2 | 5 - 6 | 1036 | 71 | |
| XM_005695599.3 | XP_005695656.2 | 5 - 6 | 993 | 71 | ||
| XM_018038354.1 | XP_017893843.1 | 3 - 4 | 262 | 71 | ||
| GenBank mRNA | KA316989.1 | - | 2 - 3 | 2562 | 71 | |
More Information
Set Membership: |
|

ZNF445
Genomic Map