Assay Details
| Gene Symbol: CRELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695620.2 | XP_005695677.2 | 7 - 8 | 975 | 54 | |
| XM_013973350.2 | XP_013828804.1 | 7 - 8 | 818 | 54 | ||
| XM_018066843.1 | XP_017922332.1 | 7 - 8 | 983 | 54 | ||
More Information
Set Membership: |
|

CRELD1
Genomic Map