Assay Details
| Gene Symbol: TWF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047903.1 | XP_017903392.1 | 5 - 6 | 689 | 76 | |
| GenBank mRNA | JO437995.2 | - | 4 - 5 | 667 | 76 | |
| JT757218.1 | - | 4 - 5 | 478 | 76 | ||
More Information
Set Membership: |
|

TWF1
Genomic Map