Assay Details
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052221.1 | XP_017907710.1 | 8 - 9 | 1950 | 81 | |
| XM_018052228.1 | XP_017907717.1 | 7 - 8 | 1778 | 81 | ||
| XM_018052237.1 | XP_017907726.1 | 7 - 8 | 1833 | 81 | ||
| XM_018052240.1 | XP_017907729.1 | 6 - 7 | 1660 | 81 | ||
| XM_018052247.1 | XP_017907736.1 | 6 - 7 | 1632 | 81 | ||
| GenBank mRNA | JO585382.2 | - | 4 - 5 | 1125 | 81 | |
| JO638640.2 | - | 1 - 2 | 792 | 81 | ||
More Information
Set Membership: |
|

SLC19A1
Genomic Map