Assay Details
| Gene Symbol: ONECUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054224.1 | XP_017909713.1 | 1 - 2 | 1359 | 64 | |
| GenBank mRNA | JO409761.2 | - | 1 - 2 | 723 | 64 | |
More Information
Set Membership: |
|

ONECUT1
Genomic Map