Assay Details
| Gene Symbol: PLEKHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969776.2 | XP_013825230.1 | 4 - 5 | 487 | 59 | |
| XM_018059368.1 | XP_017914857.1 | 4 - 5 | 630 | 59 | ||
| XM_018059369.1 | XP_017914858.1 | 4 - 5 | 630 | 59 | ||
| XM_018059370.1 | XP_017914859.1 | 4 - 5 | 486 | 59 | ||
| GenBank mRNA | JO631710.2 | - | 3 - 4 | 1408 | 59 | |
More Information
Set Membership: |
|

PLEKHB1
Genomic Map