Assay Details
| Gene Symbol: ENAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060211.1 | XP_017915700.1 | 9 - 10 | 2350 | 85 | |
| XM_018060212.1 | XP_017915701.1 | 8 - 9 | 2338 | 85 | ||
| XM_018060213.1 | XP_017915702.1 | 9 - 10 | 2349 | 85 | ||
| XM_018060214.1 | XP_017915703.1 | 7 - 8 | 2278 | 85 | ||
| XM_018060215.1 | XP_017915704.1 | 8 - 9 | 2337 | 85 | ||
| XM_018060216.1 | XP_017915705.1 | 7 - 8 | 2277 | 85 | ||
| XM_018060217.1 | XP_017915706.1 | 9 - 10 | 1649 | 85 | ||
| XM_018060218.1 | XP_017915707.1 | 9 - 10 | 1648 | 85 | ||
| XM_018060220.1 | XP_017915709.1 | 7 - 8 | 1574 | 85 | ||
| XM_018060221.1 | XP_017915710.1 | 7 - 8 | 1572 | 85 | ||
More Information
Set Membership: |
|

ENAH
Genomic Map