Assay Details
| Gene Symbol: MYLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061977.1 | XP_017917466.1 | 3 - 4 | 914 | 61 | |
| XM_018061978.1 | XP_017917467.1 | 3 - 4 | 2314 | 61 | ||
| XM_018061979.1 | XP_017917468.1 | 3 - 4 | 526 | 61 | ||
| GenBank mRNA | JO393505.2 | - | 9 - 10 | 1304 | 61 | |
More Information
Set Membership: |
|

MYLK3
Genomic Map