Assay Details
| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973342.2 | XP_013828796.2 | 1 - 2 | 2796 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPM2AIP1| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973342.2 | XP_013828796.2 | 1 - 2 | 2796 | 80 | |
Set Membership: |
|