Assay Details
| Gene Symbol: SLC35B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038773.1 | XP_017894262.1 | 3 - 4 | 482 | 83 | |
| XM_018038774.1 | XP_017894263.1 | 3 - 4 | 447 | 83 | ||
| GenBank mRNA | JO498085.2 | - | 3 - 4 | 428 | 83 | |
| JT674809.1 | - | 3 - 4 | 1395 | 83 | ||
More Information
Set Membership: |
|

SLC35B2
Genomic Map