Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038579.1 | XP_017894068.1 | 4 - 5 | 450 | 63 | |
| XM_018038580.1 | XP_017894069.1 | 4 - 5 | 447 | 63 | ||
More Information
Set Membership: |
|

KIFC1
Genomic Map