Assay Details
| Gene Symbol: GLP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039225.1 | XP_017894714.1 | 12 - 13 | 1252 | 59 | |
| GenBank mRNA | JO445664.2 | - | 7 - 8 | 605 | 59 | |
More Information
Set Membership: |
|

GLP1R
Genomic Map