Assay Details
| Gene Symbol: MYLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039259.1 | XP_017894748.1 | 12 - 13 | 1908 | 67 | |
| XM_018039260.1 | XP_017894749.1 | 12 - 13 | 1355 | 67 | ||
| XM_018039261.1 | XP_017894750.1 | 11 - 12 | 1456 | 67 | ||
| XM_018039262.1 | XP_017894751.1 | 12 - 13 | 1306 | 67 | ||
| GenBank mRNA | JO348333.2 | - | 12 - 13 | 1421 | 67 | |
| JO410852.2 | - | 12 - 13 | 1315 | 67 | ||
| JO410853.2 | - | 12 - 13 | 1315 | 67 | ||
More Information
Set Membership: |
|

MYLK4
Genomic Map