Assay Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674954.3 | XP_005675011.1 | 7 - 8 | 1510 | 63 | |
| XM_018041693.1 | XP_017897182.1 | 7 - 8 | 1281 | 63 | ||
| GenBank mRNA | JO336907.2 | - | 6 - 7 | 1053 | 63 | |
| JO357321.2 | - | 2 - 3 | 396 | 63 | ||
| JO394871.2 | - | 2 - 3 | 312 | 63 | ||
| JO415274.2 | - | 2 - 3 | 344 | 63 | ||
| JO438277.2 | - | 5 - 6 | 826 | 63 | ||
| JT762511.1 | - | 2 - 3 | 388 | 63 | ||
| KA341903.1 | - | 7 - 8 | 1487 | 63 | ||
More Information
Set Membership: |
|

TMEM39A
Genomic Map