Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675410.3 | XP_005675467.1 | 5 - 6 | 1939 | 65 | |
| GenBank mRNA | JT717112.1 | - | 6 - 7 | 1938 | 65 | |
More Information
Set Membership: |
|

SLC33A1
Genomic Map