Assay Details
| Gene Symbol: LOC102184403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679558.3 | XP_005679615.1 | 6 - 7 | 2313 | 64 | |
| XM_013963556.2 | XP_013819010.1 | 6 - 7 | 2541 | 64 | ||
| XM_013963557.2 | XP_013819011.1 | 7 - 8 | 2601 | 64 | ||
| XM_013963559.2 | XP_013819013.1 | 6 - 7 | 2313 | 64 | ||
| XM_018046832.1 | XP_017902321.1 | 6 - 7 | 2313 | 64 | ||
| XM_018046833.1 | XP_017902322.1 | 6 - 7 | 2319 | 64 | ||
| XM_018046835.1 | XP_017902324.1 | 6 - 7 | 2319 | 64 | ||
| GenBank mRNA | JO608041.2 | - | 3 - 4 | 708 | 64 | |
| JO645039.2 | - | 3 - 4 | 829 | 64 | ||
More Information
Set Membership: |
|

LOC102184403
Genomic Map