Assay Details
| Gene Symbol: LOC102184693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679940.3 | XP_005679997.1 | 2 - 3 | 697 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102184693| Gene Symbol: LOC102184693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679940.3 | XP_005679997.1 | 2 - 3 | 697 | 69 | |
Set Membership: |
|