Assay Details
| Gene Symbol: C1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680922.3 | XP_005680979.1 | 4 - 5 | 730 | 55 | |
| XM_013964253.2 | XP_013819707.1 | 4 - 5 | 758 | 55 | ||
More Information
Set Membership: |
|

C1S
Genomic Map