Assay Details
| Gene Symbol: KLHL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688864.3 | XP_005688921.2 | 2 - 3 | 1932 | 67 | |
| GenBank mRNA | JO363143.2 | - | 1 - 2 | 269 | 67 | |
More Information
Set Membership: |
|

KLHL38
Genomic Map