Assay Details
| Gene Symbol: LOC102188159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690773.3 | XP_005690830.1 | 7 - 8 | 1476 | 97 | |
| XM_018060501.1 | XP_017915990.1 | 7 - 8 | 1286 | 97 | ||
| XM_018060502.1 | XP_017915991.1 | 7 - 8 | 1309 | 97 | ||
| XM_018060503.1 | XP_017915992.1 | 7 - 8 | 1288 | 97 | ||
| XM_018060504.1 | XP_017915993.1 | 4 - 5 | 454 | 97 | ||
| GenBank mRNA | JO623761.2 | - | 3 - 4 | 1360 | 97 | |
| KA321320.1 | - | 8 - 9 | 1275 | 97 | ||
More Information
Set Membership: |
|

LOC102188159
Genomic Map