Assay Details
| Gene Symbol: FGF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692688.3 | XP_005692745.1 | 1 - 2 | 945 | 71 | |
| GenBank mRNA | KA353246.1 | - | 1 - 2 | 500 | 71 | |
| KT288195.1 | - | 1 - 2 | 247 | 71 | ||
More Information
Set Membership: |
|

FGF21
Genomic Map