Assay Details
| Gene Symbol: SLFN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693334.2 | XP_005693391.2 | 3 - 4 | 1920 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLFN14| Gene Symbol: SLFN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693334.2 | XP_005693391.2 | 3 - 4 | 1920 | 81 | |
Set Membership: |
|