Assay Details
| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693503.2 | XP_005693560.2 | 4 - 5 | 424 | 60 | |
| XM_013972198.2 | XP_013827652.2 | 4 - 5 | 409 | 60 | ||
| XM_013972199.2 | XP_013827653.1 | 4 - 5 | 409 | 60 | ||
| XM_018064583.1 | XP_017920072.1 | 4 - 5 | 409 | 60 | ||
| GenBank mRNA | JT747035.1 | - | 5 - 6 | 349 | 60 | |
| KA311468.1 | - | 4 - 5 | 317 | 60 | ||
More Information
Set Membership: |
|

CHRNB1
Genomic Map