Assay Details
| Gene Symbol: LOC102185505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694595.3 | XP_005694652.2 | 2 - 3 | 120 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102185505| Gene Symbol: LOC102185505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694595.3 | XP_005694652.2 | 2 - 3 | 120 | 127 | |
Set Membership: |
|