Assay Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695120.2 | XP_005695177.2 | 2 - 3 | 554 | 66 | |
| XM_018066121.1 | XP_017921610.1 | 2 - 3 | 256 | 66 | ||
| XM_018066122.1 | XP_017921611.1 | 2 - 3 | 245 | 66 | ||
| GenBank mRNA | JT673305.1 | - | 3 - 4 | 820 | 66 | |
More Information
Set Membership: |
|

SNUPN
Genomic Map