Assay Details
| Gene Symbol: LOC102188365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039255.1 | XP_017894744.1 | 25 - 26 | 3898 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102188365| Gene Symbol: LOC102188365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039255.1 | XP_017894744.1 | 25 - 26 | 3898 | 83 | |
Set Membership: |
|