Assay Details
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697349.3 | XP_005697406.2 | 2 - 3 | 424 | 59 | |
| XM_018041015.1 | XP_017896504.1 | 1 - 2 | 1169 | 59 | ||
More Information
Set Membership: |
|

GNG13
Genomic Map