Assay Details
| Gene Symbol: LSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698830.3 | XP_005698887.1 | 1 - 2 | 437 | 70 | |
| GenBank mRNA | KA311777.1 | - | 1 - 2 | 424 | 70 | |
More Information
Set Membership: |
|

LSM1
Genomic Map