Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699038.3 | XP_005699095.1 | 5 - 6 | 301 | 63 | |
| XM_013975553.2 | XP_013831007.1 | 5 - 6 | 277 | 63 | ||
| GenBank mRNA | JO458816.2 | - | 5 - 6 | 265 | 63 | |
More Information
Set Membership: |
|

FXYD4
Genomic Map