Assay Details
| Gene Symbol: LOC108634345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043926.1 | XP_017899415.1 | 5 - 6 | 640 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108634345| Gene Symbol: LOC108634345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043926.1 | XP_017899415.1 | 5 - 6 | 640 | 79 | |
Set Membership: |
|