Assay Details
| Gene Symbol: MCF2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047459.1 | XP_017902948.1 | 22 - 23 | 3053 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCF2L2| Gene Symbol: MCF2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047459.1 | XP_017902948.1 | 22 - 23 | 3053 | 65 | |
Set Membership: |
|